Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP2
Entrez Gene ID
102145501
Gene Name
bone morphogenetic protein 2
Gene Aliases
-
Chromosome Location
Chr.10: 55700304 - 55712467 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005568382.2 XP_005568439.1 1 - 2 771 87

View Full Product Details