Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C5AR1
Entrez Gene ID
102122506
Gene Name
complement C5a receptor 1
Gene Aliases
-
Chromosome Location
Chr.19: 48103472 - 48123279 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589697.2 XP_005589754.1 2 - 2 1545 97

View Full Product Details