Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FZD7
Entrez Gene ID
102128172
Gene Name
frizzled class receptor 7
Gene Aliases
-
Chromosome Location
Chr.12: 91461721 - 91465670 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005573955.2 XP_005574012.1 1 - 1 3148 99

View Full Product Details