Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM21
Entrez Gene ID
102119524
Gene Name
ADAM metallopeptidase domain 21
Gene Aliases
-
Chromosome Location
Chr.7: 135056983 - 135130670 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005561657.2 XP_005561714.1 2 - 2 1262 102

View Full Product Details