Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APLNR
Entrez Gene ID
102144317
Gene Name
apelin receptor
Gene Aliases
-
Chromosome Location
Chr.14: 16386286 - 16390143 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577873.2 XP_005577930.1 1 - 1 1088 116

View Full Product Details