Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTLL10
Entrez Gene ID
101926621
Gene Name
tubulin tyrosine ligase like 10
Gene Aliases
-
Chromosome Location
Chr.1: 227217455 - 227248589 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005545127.2 XP_005545184.1 12 - 13 1760 86
XM_015444135.1 XP_015299621.1 12 - 13 1816 86
XM_015444146.1 XP_015299632.1 11 - 12 2285 86
GenBank mRNA AB168848.1 - 9 - 10 1241 86

View Full Product Details