Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK17
Entrez Gene ID
102117244
Gene Name
potassium two pore domain channel subfamily K member 17
Gene Aliases
-
Chromosome Location
Chr.4: 131395025 - 131410480 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005553107.2 XP_005553164.1 2 - 3 666 69
XM_015449454.1 XP_015304940.1 2 - 3 666 69

View Full Product Details