Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN2
Entrez Gene ID
102135699
Gene Name
MORN repeat containing 2
Gene Aliases
-
Chromosome Location
Chr.13: 71788978 - 71794620 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005576065.2 XP_005576122.1 1 - 2 143 95

View Full Product Details