Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VRTN
Entrez Gene ID
102125279
Gene Name
vertebrae development associated
Gene Aliases
-
Chromosome Location
Chr.7: 139010487 - 139050662 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005561762.2 XP_005561819.1 5 - 5 2315 83
XM_015453997.1 XP_015309483.1 3 - 3 2258 83

View Full Product Details