Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC10A1
Entrez Gene ID
102138916
Gene Name
solute carrier family 10 member 1
Gene Aliases
-
Chromosome Location
Chr.7: 134442766 - 134458721 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283323.1 NP_001270252.1 1 - 2 488 76
GenBank mRNA AK240619.1 - 1 - 2 456 76
AK240620.1 - 1 - 2 489 76
KP453714.1 - 1 - 2 354 76

View Full Product Details