Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AAR2
Entrez Gene ID
101865905
Gene Name
AAR2 splicing factor
Gene Aliases
-
Chromosome Location
Chr.10: 67746529 - 67777916 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284783.1 NP_001271712.1 2 - 3 843 117
XM_005568911.2 XP_005568968.1 2 - 3 1198 117
XM_005568912.2 XP_005568969.1 2 - 3 1075 117
XM_005568913.2 XP_005568970.1 2 - 3 1096 117
XM_015457628.1 XP_015313114.1 2 - 3 1012 117
GenBank mRNA AB168889.1 - 2 - 3 843 117

View Full Product Details