The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285378.1 | NP_001272307.1 | 2 - 3 | 290 | 60 |
GenBank mRNA | AB179304.1 | - | 2 - 3 | 290 | 60 |