Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIDA
Entrez Gene ID
101925904
Gene Name
axin interactor, dorsalization associated
Gene Aliases
-
Chromosome Location
Chr.1: 81508817 - 81551817 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283182.1 NP_001270111.1 4 - 5 847 101
XM_015440188.1 XP_015295674.1 5 - 6 557 101
GenBank mRNA AB168527.1 - 4 - 5 847 101

View Full Product Details