Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD9A
Entrez Gene ID
101926088
Gene Name
RAD9 checkpoint clamp component A
Gene Aliases
-
Chromosome Location
Chr.14: 7055383 - 7142517 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284976.1 NP_001271905.1 7 - 8 748 97
XM_015434151.1 XP_015289637.1 2 - 3 637 97
XM_015434152.1 XP_015289638.1 7 - 8 1982 97
XM_015434153.1 XP_015289639.1 7 - 8 2262 97
XM_015434154.1 XP_015289640.1 7 - 8 1687 97
XM_015434155.1 XP_015289641.1 6 - 7 1256 97
XM_015434156.1 XP_015289642.1 6 - 7 1151 97
XM_015434157.1 XP_015289643.1 2 - 3 637 97
XM_015434158.1 XP_015289644.1 7 - 8 1975 97
XM_015434159.1 XP_015289645.1 2 - 3 595 97
XM_015434160.1 XP_015289646.1 7 - 8 1937 97
XM_015434161.1 XP_015289647.1 6 - 7 1910 97
GenBank mRNA AB169413.1 - 7 - 8 748 97

View Full Product Details