Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAD9A
Entrez Gene ID
101926088
Gene Name
RAD9 checkpoint clamp component A
Gene Aliases
-
Chromosome Location
Chr.14: 7055383 - 7142517 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284976.1 NP_001271905.1 8 - 9 849 89
XM_015434151.1 XP_015289637.1 3 - 4 738 89
XM_015434152.1 XP_015289638.1 8 - 9 2083 89
XM_015434153.1 XP_015289639.1 8 - 9 2363 89
XM_015434154.1 XP_015289640.1 8 - 9 1788 89
XM_015434155.1 XP_015289641.1 7 - 8 1357 89
XM_015434156.1 XP_015289642.1 7 - 8 1252 89
XM_015434157.1 XP_015289643.1 3 - 4 738 89
XM_015434158.1 XP_015289644.1 8 - 9 2076 89
XM_015434159.1 XP_015289645.1 3 - 4 696 89
XM_015434160.1 XP_015289646.1 8 - 9 2038 89
XM_015434161.1 XP_015289647.1 7 - 8 2011 89
GenBank mRNA AB169413.1 - 8 - 9 849 89

View Full Product Details