Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQUB
Entrez Gene ID
101926148
Gene Name
IQ motif and ubiquitin domain containing
Gene Aliases
QtsA-17428
Chromosome Location
Chr.3: 156254036 - 156336527 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283096.1 NP_001270025.1 2 - 3 482 73
XM_005550638.2 XP_005550695.1 3 - 4 1011 73
XM_005550639.2 XP_005550696.1 2 - 3 974 73
XM_005550640.2 XP_005550697.1 2 - 3 980 73
XM_015447303.1 XP_015302789.1 2 - 3 417 73
GenBank mRNA AB169093.1 - 2 - 3 482 73

View Full Product Details