Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQUB
Entrez Gene ID
101926148
Gene Name
IQ motif and ubiquitin domain containing
Gene Aliases
QtsA-17428
Chromosome Location
Chr.3: 156254036 - 156336527 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283096.1 NP_001270025.1 9 - 10 1665 96
XM_005550638.2 XP_005550695.1 10 - 11 2194 96
XM_005550639.2 XP_005550696.1 8 - 9 1995 96
XM_015447303.1 XP_015302789.1 9 - 10 1600 96
GenBank mRNA AB169093.1 - 9 - 10 1665 96

View Full Product Details