Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEKT5
Entrez Gene ID
101926637
Gene Name
tektin 5
Gene Aliases
-
Chromosome Location
Chr.20: 10903501 - 10971435 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285262.1 NP_001272191.1 1 - 2 606 63
GenBank mRNA AB179415.1 - 1 - 2 606 63

View Full Product Details