The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001283713.1 | NP_001270642.1 | 2 - 3 | 1213 | 105 |
GenBank mRNA | AB170778.1 | - | 2 - 3 | 1213 | 105 |