Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKR1C1
Entrez Gene ID
101865536
Gene Name
Aldo-keto reductase family 1 member C1 homolog
Gene Aliases
DD1, dd-1
Chromosome Location
Chr.9: 5321817 - 5336693 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283533.1 NP_001270462.1 9 - 9 1220 133
GenBank mRNA AB174272.1 - 9 - 9 1220 133

View Full Product Details