Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMACR
Entrez Gene ID
102130505
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
-
Chromosome Location
Chr.6: 34719362 - 34742780 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005556674.2 XP_005556731.1 3 - 4 657 96

View Full Product Details