Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALR3
Entrez Gene ID
102123886
Gene Name
galanin receptor 3
Gene Aliases
-
Chromosome Location
Chr.10: 13417125 - 13420781 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015456974.1 XP_015312460.1 1 - 2 1824 122

View Full Product Details