Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPPA3
Entrez Gene ID
102146769
Gene Name
developmental pluripotency associated 3
Gene Aliases
-
Chromosome Location
Chr.11: 8126680 - 8134631 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570022.2 XP_005570079.1 3 - 4 776 86

View Full Product Details