Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL23
Entrez Gene ID
102136663
Gene Name
methyltransferase like 23
Gene Aliases
-
Chromosome Location
Chr.16: 74108826 - 74114989 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015438886.1 XP_015294372.1 3 - 4 760 91
XM_015438888.1 XP_015294374.1 3 - 4 409 91

View Full Product Details