Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCHE
Entrez Gene ID
102130167
Gene Name
butyrylcholinesterase
Gene Aliases
-
Chromosome Location
Chr.2: 69517015 - 69585445 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319370.1 NP_001306299.1 1 - 2 1523 80
XM_005546296.2 XP_005546353.1 2 - 3 1684 80
GenBank mRNA GU565682.1 - 1 - 2 1523 80

View Full Product Details