Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL17A
Entrez Gene ID
102119976
Gene Name
interleukin 17A
Gene Aliases
-
Chromosome Location
Chr.4: 118459415 - 118462934 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552759.2 XP_005552816.1 2 - 3 278 81

View Full Product Details