Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKX3-2
Entrez Gene ID
102115367
Gene Name
NK3 homeobox 2
Gene Aliases
-
Chromosome Location
Chr.5: 12624048 - 12627725 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005554495.2 XP_005554552.1 1 - 2 577 63

View Full Product Details