Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRIT3
Entrez Gene ID
102118735
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 3
Gene Aliases
-
Chromosome Location
Chr.5: 109224723 - 109250242 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555666.2 XP_005555723.1 2 - 3 816 65

View Full Product Details