Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAGP
Entrez Gene ID
102136717
Gene Name
small cell adhesion glycoprotein
Gene Aliases
-
Chromosome Location
Chr.11: 50151449 - 50177108 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570880.2 XP_005570937.1 2 - 3 267 62

View Full Product Details