Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUXA
Entrez Gene ID
102132503
Gene Name
double homeobox A
Gene Aliases
-
Chromosome Location
Chr.19: 57828847 - 57837149 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590495.1 XP_005590552.1 4 - 5 439 109

View Full Product Details