Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102122124
Entrez Gene ID
102122124
Gene Name
nuclear pore complex-interacting protein family member B11-like
Gene Aliases
-
Chromosome Location
Chr.20: 15149753 - 15170922 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591304.2 XP_005591361.1 8 - 9 692 70
XM_015442330.1 XP_015297816.1 8 - 9 741 70

View Full Product Details