Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DELEC1
Entrez Gene ID
102117078
Gene Name
deleted in esophageal cancer 1
Gene Aliases
DEC1
Chromosome Location
Chr.15: 22070083 - 22077606 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005582355.1 XP_005582412.1 3 - 4 287 103

View Full Product Details