Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IBA57
Entrez Gene ID
102137147
Gene Name
iron-sulfur cluster assembly factor IBA57
Gene Aliases
-
Chromosome Location
Chr.1: 87023803 - 87030082 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015450898.1 XP_015306384.1 1 - 2 4105 65
XM_015450905.1 XP_015306391.1 2 - 3 892 65
XM_015450913.1 XP_015306399.1 2 - 3 420 65

View Full Product Details