Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HES5
Entrez Gene ID
102143983
Gene Name
hes family bHLH transcription factor 5
Gene Aliases
-
Chromosome Location
Chr.1: 225846648 - 225847806 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005545236.2 XP_005545293.1 1 - 2 149 68

View Full Product Details