Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCLK3
Entrez Gene ID
102131340
Gene Name
doublecortin like kinase 3
Gene Aliases
-
Chromosome Location
Chr.2: 91559850 - 91584657 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005546620.2 XP_005546677.1 2 - 3 1934 100

View Full Product Details