Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRE3
Entrez Gene ID
102118304
Gene Name
adhesion G protein-coupled receptor E3
Gene Aliases
EMR3
Chromosome Location
Chr.19: 15011547 - 15079187 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015440203.1 XP_015295689.1 6 - 7 1080 102

View Full Product Details