Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERICH5
Entrez Gene ID
102126957
Gene Name
glutamate rich 5
Gene Aliases
C8H8orf47
Chromosome Location
Chr.8: 99215269 - 99243833 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563769.2 XP_005563826.1 1 - 2 524 88

View Full Product Details