Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM72
Entrez Gene ID
102126840
Gene Name
transmembrane protein 72
Gene Aliases
-
Chromosome Location
Chr.9: 43556944 - 43582229 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565098.2 XP_005565155.1 3 - 4 550 64

View Full Product Details