Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP35
Entrez Gene ID
102130204
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 35
Gene Aliases
-
Chromosome Location
Chr.11: 127769068 - 127776980 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005572563.2 XP_005572620.1 1 - 2 721 64

View Full Product Details