Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO39
Entrez Gene ID
102131314
Gene Name
F-box protein 39
Gene Aliases
-
Chromosome Location
Chr.16: 6794303 - 6804888 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005582678.2 XP_005582735.1 2 - 3 1212 73
XM_005582680.2 XP_005582737.1 2 - 3 1193 73

View Full Product Details