Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VGLL1
Entrez Gene ID
102137749
Gene Name
vestigial like family member 1
Gene Aliases
-
Chromosome Location
Chr.X: 133299994 - 133336438 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015444336.1 XP_015299822.1 3 - 4 738 74
XM_015444337.1 XP_015299823.1 2 - 3 649 74

View Full Product Details