Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP57
Entrez Gene ID
102121260
Gene Name
ZFP57 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.4: 141152207 - 141157527 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005595976.2 XP_005596033.2 3 - 4 1295 92

View Full Product Details