Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXA6
Entrez Gene ID
102138229
Gene Name
homeobox A6
Gene Aliases
-
Chromosome Location
Chr.3: 93580603 - 93582771 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549936.2 XP_005549993.1 1 - 2 463 84

View Full Product Details