Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102129273
Entrez Gene ID
102129273
Gene Name
5-hydroxytryptamine receptor 3E
Gene Aliases
HTR3E
Chromosome Location
Chr.2: 88779986 - 88789067 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005595957.2 XP_005596014.1 6 - 7 768 86

View Full Product Details