Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XRCC2
Entrez Gene ID
102133387
Gene Name
X-ray repair cross complementing 2
Gene Aliases
-
Chromosome Location
Chr.3: 185358985 - 185392433 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005551231.2 XP_005551288.1 1 - 2 167 108

View Full Product Details