Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHNAK2
Entrez Gene ID
102145969
Gene Name
AHNAK nucleoprotein 2
Gene Aliases
-
Chromosome Location
Chr.7: 170315563 - 170342036 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015454263.1 XP_015309749.1 6 - 7 1053 73

View Full Product Details