Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC10B
Entrez Gene ID
102136105
Gene Name
leucine rich repeat containing 10B
Gene Aliases
-
Chromosome Location
Chr.14: 12753044 - 12755046 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005578864.1 XP_005578921.1 1 - 1 723 87

View Full Product Details