Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF16
Entrez Gene ID
102118909
Gene Name
zinc finger protein 16
Gene Aliases
-
Chromosome Location
Chr.8: 146631765 - 146649291 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005564430.2 XP_005564487.1 2 - 3 431 58
XM_005564433.2 XP_005564490.1 2 - 3 1708 58
XM_005564434.2 XP_005564491.1 2 - 3 1691 58
XM_005564435.2 XP_005564492.1 2 - 3 300 58

View Full Product Details