Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SBSN
Entrez Gene ID
102138840
Gene Name
suprabasin
Gene Aliases
-
Chromosome Location
Chr.19: 35972568 - 35982573 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005588865.2 XP_005588922.1 2 - 3 3664 79
XM_015441696.1 XP_015297182.1 3 - 4 2710 79

View Full Product Details