Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FILIP1L
Entrez Gene ID
102141662
Gene Name
filamin A interacting protein 1 like
Gene Aliases
-
Chromosome Location
Chr.2: 172034727 - 172329127 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005548313.2 XP_005548370.1 2 - 3 711 98

View Full Product Details